Gene Transcripts

Gene Symbol
Lcat
Entrez Gene ID
16816
Gene Name
lecithin cholesterol acyltransferase
Gene Aliases
AI046659, D8Wsu61e
Chromosome Location
Chr. 8: 105939551 - 105943402 on Build GRCm38
UniGene ID
Mm.1593
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_008490.2 NP_032516.2 5 595
GenBank mRNA AI574112.1 - 3 335
AK004914.1 - 5 591
AK149476.1 - 5 592
BC028861.1 - 5 578 4212194
J05154.1 - 5 575

View Full Product Details