Gene Transcripts

Gene Symbol
Tm4sf1
Entrez Gene ID
17112
Gene Name
transmembrane 4 superfamily member 1
Gene Aliases
L6, M3s1
Chromosome Location
Chr. 3: 57287064 - 57301983 on Build GRCm38
UniGene ID
Mm.856
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_008536.3 NP_032562.1 6 1051
GenBank mRNA AK077326.1 - 4 554
AK133556.1 - 552
AK134603.1 - 5 987
AK164718.1 - 4 555
BC083073.1 - 6 1051 6337376
KC508089.1 - 4 526
L15429.1 - 4 541
L15443.1 - 4 541

View Full Product Details