Gene Transcripts
- Gene Symbol
- Pax6
- Entrez Gene ID
- 18508
- Gene Name
- paired box 6
- Gene Aliases
- 1500038E17Rik, AEY11, Dey, Gsfaey11, Pax-6, Sey
- Chromosome Location
- Chr. 2: 105667518 - 105697364 on Build GRCm38
- UniGene ID
- Mm.33870
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001244198.1 |
NP_001231127.1
|
10, 11 |
1244 |
|
NM_001244200.1 |
NP_001231129.1
|
11, 12 |
1456 |
|
NM_001244201.1 |
NP_001231130.1
|
9, 10 |
1202 |
|
NM_001244202.1 |
NP_001231131.1
|
9, 10 |
1758 |
|
NM_013627.5 |
NP_038655.1
|
10, 11 |
1800 |
|
GenBank mRNA |
AF443223.1 |
- |
6, 7 |
857 |
|
AF457141.1 |
- |
4, 5 |
523 |
|
AJ292077.1 |
- |
9, 10 |
1061 |
|
AJ307468.1 |
- |
9, 10 |
1068 |
|
AK028059.1 |
- |
9, 10 |
1203 |
|
AK032616.1 |
- |
10, 11 |
1245 |
|
AK045805.1 |
- |
9, 10 |
1760 |
|
AK139054.1 |
- |
|
788 |
|
AY064175.1 |
- |
5, 6 |
667 |
|
BC011272.1 |
- |
10, 11 |
1488 |
4008490 |
BC036957.1 |
- |
9, 10 |
1198 |
4504106 |
BC069912.1 |
- |
6, 7 |
1235 |
|
M77842.1 |
- |
5, 6 |
787 |
|
X63963.1 |
- |
10, 11 |
1121 |
|
View Full Product Details