Gene Transcripts

Gene Symbol
Trpv5
Entrez Gene ID
194352
Gene Name
transient receptor potential cation channel, subfamily V, member 5
Gene Aliases
CAT2, D630033B11, ECAC1
Chromosome Location
Chr. 6: 41651661 - 41682393 on Build GRCm38
UniGene ID
Mm.135734
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001007572.2 NP_001007573.1 8 1152
GenBank mRNA AK085479.1 - 8 1198
BC110554.1 - 8 1152 40047811
BC110555.1 - 8 1152 40047816

View Full Product Details