Gene Transcripts
- Gene Symbol
- Sh2b1
- Entrez Gene ID
- 20399
- Gene Name
- SH2B adaptor protein 1
- Gene Aliases
- AI425885, C530001K22Rik, Irip, Psm, SH2-B, SH2-Bb, Sh2bpsm1, mKIAA1299
- Chromosome Location
- Chr. 7: 126466993 - 126475684 on Build GRCm38
- UniGene ID
- Mm.8538
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001081459.2 |
NP_001074928.1
|
7, 8 |
2326 |
|
NM_001289538.1 |
NP_001276467.1
|
7, 8 |
2326 |
|
NM_001289539.1 |
NP_001276468.1
|
7, 8 |
1949 |
|
NM_001289540.1 |
NP_001276469.1
|
8, 9 |
2086 |
|
NM_001289541.1 |
NP_001276470.1
|
7, 8 |
1949 |
|
NM_001289542.1 |
NP_001276471.1
|
7, 8 |
2326 |
|
NM_011363.3 |
NP_035493.2
|
7, 8 |
2326 |
|
GenBank mRNA |
AF020526.1 |
- |
7, 8 |
2020 |
|
AF036355.1 |
- |
1, 2 |
182 |
|
AF074329.1 |
- |
6, 7 |
1671 |
|
AF380422.1 |
- |
7, 8 |
2020 |
|
AF421138.1 |
- |
7, 8 |
2020 |
|
AF421139.1 |
- |
7, 8 |
2020 |
|
AK148085.1 |
- |
8, 9 |
2053 |
|
AK154655.1 |
- |
7, 8 |
2260 |
|
AK168439.1 |
- |
7, 8 |
1921 |
|
AK170444.1 |
- |
7, 8 |
2300 |
|
AK171328.1 |
- |
7, 8 |
2334 |
|
AK198191.1 |
- |
2, 3 |
350 |
|
BC011422.1 |
- |
7, 8 |
1871 |
3156603 |
BC051978.1 |
- |
7, 8 |
1869 |
6401205 |
View Full Product Details