Gene Transcripts

Gene Symbol
Rab40c
Entrez Gene ID
224624
Gene Name
Rab40C, member RAS oncogene family
Gene Aliases
RAR3
Chromosome Location
Chr. 17: 25882114 - 25919714 on Build GRCm38
UniGene ID
Mm.328872
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_139154.2 NP_631893.1 7 933
GenBank mRNA AB232639.1 - 6 827
AF422144.1 - 6 886
AK133766.1 - 6 709
BC027008.1 - 7 921 5364768
BC037461.1 - 8 1365

View Full Product Details