Gene Transcripts
- Gene Symbol
- B3gnt4
- Entrez Gene ID
- 231727
- Gene Name
- UDP-GlcNAc:betaGal beta-1,3-N-acetylglucosaminyltransferase 4
- Gene Aliases
- 1010001G17Rik, BGnT-4
- Chromosome Location
- Chr. 5: 123508289 - 123512099 on Build GRCm38
- UniGene ID
- Mm.484068
- Species
- Mus musculus
- Gene Symbol
- Diablo
- Entrez Gene ID
- 66593
- Gene Name
- diablo homolog (Drosophila)
- Gene Aliases
- 0610041G12Rik, 1700006L01Rik, AU040403, Smac
- Chromosome Location
- Chr. 5: 123511330 - 123524164 on Build GRCm38
- UniGene ID
- Mm.46716
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK133194.1 |
- |
|
2416 |
|
AK135627.1 |
- |
|
349 |
|
BC094617.1 |
- |
|
44 |
|
View Full Product Details