Gene Transcripts

Gene Symbol
Lrrc10
Entrez Gene ID
237560
Gene Name
leucine rich repeat containing 10
Gene Aliases
D330003I11Rik, Hrlrrp, Serdin1
Chromosome Location
Chr. 10: 117045341 - 117046768 on Build GRCm38
UniGene ID
Mm.37320
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_146242.2 NP_666354.1 1 332
GenBank mRNA AB091100.1 - 1 302
AF527781.1 - 1 318
AK084466.1 - 1 317
AK085779.1 - 1 320
AK142167.1 - 1 319
BC086911.1 - 1 332 6752714

View Full Product Details