Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001081143.1 | NP_001074612.1 | 4 | 476 | |
GenBank mRNA | AK040065.1 | - | 4 | 477 | |
BC147449.1 | - | 4 | 497 | 9056091 | |
BC147466.1 | - | 4 | 497 | 9056108 | |
BC147839.1 | - | 4 | 425 | 9087556 | |
BC147841.1 | - | 4 | 425 | 9087558 |