Gene Transcripts

Gene Symbol
Elfn1
Entrez Gene ID
243312
Gene Name
leucine rich repeat and fibronectin type III, extracellular 1
Gene Aliases
A930017N06Rik, AI197272, Ppp1r28
Chromosome Location
Chr. 5: 139907640 - 139974725 on Build GRCm38
UniGene ID
Mm.237102
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_175522.3 NP_780731.1 2 1015
GenBank mRNA AK044511.1 - 2 1016
BC059029.1 - 1 747 6811341

View Full Product Details