Gene Transcripts

Gene Symbol
Cyp2b23
Entrez Gene ID
243881
Gene Name
cytochrome P450, family 2, subfamily b, polypeptide 23
Gene Aliases
EG243881
Chromosome Location
Chr. 7: 26665227 - 26690372 on Build GRCm38
UniGene ID
Mm.294678
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001081148.1 NP_001074617.1 2 238

View Full Product Details