Gene Transcripts

Gene Symbol
Olfr77
Entrez Gene ID
258336
Gene Name
olfactory receptor 77
Gene Aliases
18A, MOR143-1
Chromosome Location
Chr. 9: 19917229 - 19922510 on Build GRCm38
UniGene ID
Mm.261577
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_146339.3 NP_666451.1 3 942
GenBank mRNA BC046590.1 - 3 942

View Full Product Details