Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_172300.3 | NP_758504.3 | 6 | 919 | |
GenBank mRNA | AK005524.1 | - | 4 | 470 | |
AK077793.1 | - | 6 | 918 | ||
AK149770.1 | - | 6 | 919 | ||
AK152529.1 | - | 6 | 633 | ||
AK153328.1 | - | 6 | 650 | ||
AK168942.1 | - | 6 | 612 | ||
AK208933.1 | - | 2 | 38 | ||
BC027266.1 | - | 6 | 544 | ||
BC054412.1 | - | 6 | 622 | 2646768 |