Gene Transcripts

Gene Symbol
Rtn4rl2
Entrez Gene ID
269295
Gene Name
reticulon 4 receptor-like 2
Gene Aliases
Ngr2, Ngrh1, Ngrl3
Chromosome Location
Chr. 2: 84871309 - 84888507 on Build GRCm38
UniGene ID
Mm.389289
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_199223.1 NP_954693.1 2 402
GenBank mRNA AY250220.1 - 2 402
BC132523.1 - 2 466 40130800
BC138154.1 - 2 466 8861173

View Full Product Details