Gene Transcripts

Gene Symbol
Psmd10
Entrez Gene ID
53380
Gene Name
proteasome (prosome, macropain) 26S subunit, non-ATPase, 10
Gene Aliases
AW554874
Chromosome Location
Chr. X: 140948425 - 140956711 on Build GRCm38
UniGene ID
Mm.17640
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001164177.1 NP_001157649.1 3 326
NM_016883.4 NP_058579.2 3 326
GenBank mRNA AB022022.1 - 3 282
AK009068.1 - 3 305
AK018233.1 - 3 304
AK136400.1 - 3 305
BC026931.1 - 3 293 3987112
BC056196.1 - 3 311 30019985
BY057263.1 - 3 326
CD564300.1 - 3 293

View Full Product Details