Gene Transcripts
- Gene Symbol
- Alyref2
- Entrez Gene ID
- 56009
- Gene Name
- Aly/REF export factor 2
- Gene Aliases
- C130042O11Rik, REF2, Refbp2
- Chromosome Location
- Chr. 1: 171503478 - 171504750 on Build GRCm38
- UniGene ID
- Mm.389208
- Species
- Mus musculus
- Gene Symbol
- B930036N10Rik
- Entrez Gene ID
- 100048917
- Gene Name
- RIKEN cDNA B930036N10 gene
- Gene Aliases
- Chromosome Location
- Chr. : - on Build
- UniGene ID
- Mm.389208
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK047216.1 |
- |
1 |
414 |
|
View Full Product Details