Gene Transcripts
- Gene Symbol
- Rab37
- Entrez Gene ID
- 58222
- Gene Name
- RAB37, member RAS oncogene family
- Gene Aliases
- B230331O03Rik, B230354I04Rik
- Chromosome Location
- Chr. 11: 115091203 - 115162773 on Build GRCm38
- UniGene ID
- Mm.143789
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK050483.1 |
- |
2, 3 |
209 |
|
View Full Product Details