Gene Transcripts

Gene Symbol
Tmem86a
Entrez Gene ID
67893
Gene Name
transmembrane protein 86A
Gene Aliases
1810054O13Rik, AI414959, AW742698
Chromosome Location
Chr. 7: 47050640 - 47054776 on Build GRCm38
UniGene ID
Mm.27338
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_026436.3 NP_080712.1 3 839
GenBank mRNA AK007864.1 - 3 840
AK075589.1 - 3 813
BC056619.1 - 3 806 4987281

View Full Product Details