Gene Transcripts

Gene Symbol
Mpnd
Entrez Gene ID
68047
Gene Name
MPN domain containing
Gene Aliases
E130307M08Rik
Chromosome Location
Chr. 17: 56009157 - 56016790 on Build GRCm38
UniGene ID
Mm.358710
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_026530.5 NP_080806.4 8, 9 1006
GenBank mRNA AK012128.1 - 7, 8 961
AK146973.1 - 8, 9 991
AK160356.1 - 8, 9 975
AK172381.1 - 7, 8 749
AK185646.1 - 3, 4 187
AK216749.1 - 2, 3 136
BC017625.1 - 7, 8 955

View Full Product Details