Gene Transcripts
- Gene Symbol
- Rbfa
- Entrez Gene ID
- 68731
- Gene Name
- ribosome binding factor A
- Gene Aliases
- 1110032A13Rik, AI595940
- Chromosome Location
- Chr. 18: 80192264 - 80200619 on Build GRCm38
- UniGene ID
- Mm.186936
- Species
- Mus musculus
- Gene Symbol
- LOC105246506
- Entrez Gene ID
- 105246506
- Gene Name
- uncharacterized LOC105246506
- Gene Aliases
- Chromosome Location
- Chr. 18: 80186234 - 80192175 on Build GRCm38
- UniGene ID
- Mm.392326
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK141205.1 |
- |
|
1946 |
|
View Full Product Details