Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001172154.1 | NP_001165625.1 | 5 | 655 | |
NM_027109.2 | NP_081385.1 | 5 | 772 | ||
GenBank mRNA | AK009174.1 | - | 5 | 778 | |
AK150242.1 | - | 5 | 685 | ||
BC023246.1 | - | 5 | 655 | 4039470 | |
DQ116784.1 | - | 5 | 666 |