Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001171002.1 | NP_001164473.1 | 2 | 383 | |
NM_027299.5 | NP_081575.2 | 2 | 383 | ||
GenBank mRNA | AF466377.1 | - | 2 | 371 | |
AK008666.1 | - | 2 | 384 | ||
AK162979.1 | - | 2 | 376 | ||
BC016427.1 | - | 2 | 365 | 4218832 | |
BC035454.1 | - | 2 | 365 |