Gene Transcripts

Gene Symbol
Aox3
Entrez Gene ID
71724
Gene Name
aldehyde oxidase 3
Gene Aliases
1200011D03Rik, AI326299, Aoh1, Aoh2
Chromosome Location
Chr. 1: 58113136 - 58200452 on Build GRCm38
UniGene ID
Mm.20108
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_023617.2 NP_076106.2 34, 35 4084
GenBank mRNA AF172276.2 - 34, 35 4163
AK004700.1 - 23, 24 2880
AK030109.1 - 30, 31 3625
AK149434.1 - 34, 35 4046
AY347569.1 - 34, 35 3964
AY347570.1 - 34, 35 3964
BC049853.1 - 34, 35 4035
BC138876.1 - 34, 35 4063 8861898

View Full Product Details