Gene Transcripts

Gene Symbol
Tmem81
Entrez Gene ID
74626
Gene Name
transmembrane protein 81
Gene Aliases
4930429O20Rik, AI465369
Chromosome Location
Chr. 1: 132506230 - 132508639 on Build GRCm38
UniGene ID
Mm.34611
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_029025.3 NP_083301.1 2 773
GenBank mRNA AK015245.1 - 2 774
AK045585.1 - 2 713
BC095954.1 - 1 786 5068880
BC120863.1 - 2 533 40129786
BC132045.1 - 2 533 40130322

View Full Product Details