Gene Transcripts

Gene Symbol
Grhpr
Entrez Gene ID
76238
Gene Name
glyoxylate reductase/hydroxypyruvate reductase
Gene Aliases
1110059D05Rik, 6430629L09Rik
Chromosome Location
Chr. 4: 44981387 - 44992085 on Build GRCm38
UniGene ID
Mm.196574
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_080289.1 NP_525028.1 2 173
GenBank mRNA AK004307.1 - 2 190
AK018293.1 - 2 189
AK168351.1 - 2 170
AK172201.1 - 2 141
AY113690.1 - 2 147
BC010194.1 - 2 173 3966541

View Full Product Details