Gene Transcripts

Gene Symbol
Mib2
Entrez Gene ID
76580
Gene Name
mindbomb homolog 2 (Drosophila)
Gene Aliases
2210008I11Rik, Zzank1, skd
Chromosome Location
Chr. 4: 155654470 - 155669254 on Build GRCm38
UniGene ID
Mm.235738
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001256107.1 NP_001243036.1 3 1081
NM_001256108.2 NP_001243037.2 4 997
NM_145124.3 NP_660106.2 3 1081
GenBank mRNA AB063290.1 - 1 84
AB072336.1 - 3 643
AK147940.1 - 4 941
AK159611.1 - 3 799
AK181495.1 - 1 42
AY974090.1 - 1 222
BC058086.1 - 2 340
BC141133.1 - 2 395 9055745
BC145441.1 - 2 395

View Full Product Details