Gene Transcripts
- Gene Symbol
- Atg16l1
- Entrez Gene ID
- 77040
- Gene Name
- autophagy related 16-like 1 (S. cerevisiae)
- Gene Aliases
- 1500009K01Rik, Apg16l, Atg16l, WDR30
- Chromosome Location
- Chr. 1: 87756006 - 87792428 on Build GRCm38
- UniGene ID
- Mm.272972
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001205391.1 |
NP_001192320.1
|
16, 17 |
1819 |
|
NM_001205392.1 |
NP_001192321.1
|
14, 15 |
1714 |
|
NM_029846.4 |
NP_084122.2
|
15, 16 |
1771 |
|
GenBank mRNA |
AB087879.1 |
- |
14, 15 |
1525 |
|
AB087880.1 |
- |
15, 16 |
1582 |
|
AB087881.1 |
- |
16, 17 |
1630 |
|
AB476646.1 |
- |
16, 17 |
1630 |
|
AK005181.1 |
- |
15, 16 |
1988 |
|
AK030983.1 |
- |
15, 16 |
1712 |
|
AK131120.1 |
- |
13, 14 |
2801 |
|
AK155358.1 |
- |
14, 15 |
1650 |
|
AK170073.1 |
- |
14, 15 |
1648 |
|
BC049122.1 |
- |
16, 17 |
1744 |
6813377 |
BC052087.1 |
- |
3, 4 |
669 |
|
View Full Product Details