Gene Transcripts

Gene Symbol
Vmn1r56
Entrez Gene ID
81015
Gene Name
vomeronasal 1 receptor 56
Gene Aliases
V1rd3, V3R3
Chromosome Location
Chr. 7: 5194916 - 5196747 on Build GRCm38
UniGene ID
Mm.160380
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_030740.1 NP_109665.1 1 232
GenBank mRNA AF324868.1 - 1 232

View Full Product Details