Gene Transcripts

Gene Symbol
Ankrd39
Entrez Gene ID
109346
Gene Name
ankyrin repeat domain 39
Gene Aliases
9130416N05Rik, C030004B10Rik
Chromosome Location
Chr. 1: 36538173 - 36547440 on Build GRCm38
UniGene ID
Mm.247537
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_026241.3 NP_080517.1 3 258
GenBank mRNA AK018681.1 - 3 259
AK030874.1 - 454
AK042896.1 - 3 259
AK081207.1 - 3 451
AK158215.1 - 3 453
AK160231.1 - 3 274
BC117004.1 - 3 288 40126323
BC119100.1 - 3 288 8733849
BC144893.1 - 3 288

View Full Product Details