Gene Transcripts

Gene Symbol
Htr1d
Entrez Gene ID
15552
Gene Name
5-hydroxytryptamine (serotonin) receptor 1D
Gene Aliases
5-HT-1D, 5-HT1D, AI853647, Gpcr14, Htr1db
Chromosome Location
Chr. 4: 136423522 - 136444405 on Build GRCm38
UniGene ID
Mm.40573
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001285482.1 NP_001272411.1 3 2071
NM_001285483.1 NP_001272412.1 3 1893
NM_001285484.1 NP_001272413.1 2 1800
NM_008309.5 NP_032335.2 2 1913
GenBank mRNA AK082016.1 - 2 1917

View Full Product Details