Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_010531.1 | NP_034661.1 | 6 | 570 | |
GenBank mRNA | AB019505.1 | - | 4 | 419 | |
AF110802.1 | - | 4 | 1797 | ||
AF110803.1 | - | 5 | 497 | ||
AF122907.1 | - | 6 | 570 | ||
AK003370.1 | - | 6 | 586 | ||
AK008452.1 | - | 5 | 501 | ||
AK009721.1 | - | 5 | 497 | ||
AK009877.1 | - | 2 | 137 | ||
BC018332.1 | - | 5 | 480 | 4007411 | |
CT010164.1 | - | 4 | 419 | ||
CT010334.1 | - | 4 | 419 |