Gene Transcripts

Gene Symbol
Myoz3
Entrez Gene ID
170947
Gene Name
myozenin 3
Gene Aliases
4833419K08Rik, Fatz-3
Chromosome Location
Chr. 18: 60573717 - 60591716 on Build GRCm38
UniGene ID
Mm.159257
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_133363.3 NP_579941.2 3 210
GenBank mRNA AF480442.1 - 3 288
BC104046.2 - 3 210 40040057
BC104047.1 - 3 210 40040058

View Full Product Details