Gene Transcripts

Gene Symbol
Hsh2d
Entrez Gene ID
209488
Gene Name
hematopoietic SH2 domain containing
Gene Aliases
ALX, Hsh2
Chromosome Location
Chr. 8: 72189668 - 72200958 on Build GRCm38
UniGene ID
Mm.157431
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_197944.1 NP_922935.1 6 669
GenBank mRNA AK157000.1 - 702
AY319653.1 - 6 669
BC094291.1 - 5 648 5316178

View Full Product Details