Gene Transcripts

Gene Symbol
Taar4
Entrez Gene ID
209513
Gene Name
trace amine-associated receptor 4
Gene Aliases
Gm226, Ta2, taR-4
Chromosome Location
Chr. 10: 23960494 - 23961537 on Build GRCm38
UniGene ID
Mm.300612
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001008499.1 NP_001008499.1 1 474
GenBank mRNA BC117729.1 - 1 473
BC117730.1 - 1 463

View Full Product Details