Gene Transcripts

Gene Symbol
Gxylt1
Entrez Gene ID
223827
Gene Name
glucoside xylosyltransferase 1
Gene Aliases
Glt8d3, Gm1228, Gm87
Chromosome Location
Chr. 15: 93239735 - 93275336 on Build GRCm38
UniGene ID
Mm.22819
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001033275.4 NP_001028447.1 3 462
GenBank mRNA AK147387.1 - 3 463
BC141302.1 - 3 557 9055914
BC171972.1 - 3 557 9054258

View Full Product Details