Gene Transcripts

Gene Symbol
Olfr96
Entrez Gene ID
258507
Gene Name
olfactory receptor 96
Gene Aliases
MOR121-1
Chromosome Location
Chr. 17: 37225127 - 37226068 on Build GRCm38
UniGene ID
Mm.377543
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_146514.1 NP_666725.1 1 761
GenBank mRNA BC120652.1 - 1 797

View Full Product Details