Gene Transcripts

Gene Symbol
Esrra
Entrez Gene ID
26379
Gene Name
estrogen related receptor, alpha
Gene Aliases
ERRalpha, Err1, Estrra, Nr3b1
Chromosome Location
Chr. 19: 6910977 - 6922048 on Build GRCm38
UniGene ID
Mm.386776
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_007953.2 NP_031979.2 2 413
GenBank mRNA AK019222.1 - 2 374
AK046588.1 - 2 373
AK156371.1 - 2 389
BC034165.1 - 2 356
BC039774.1 - 2 425
BC171958.1 - 2 373 9054244
CO425338.1 - 1 63
U85259.1 - 2 380

View Full Product Details