Gene Transcripts

Gene Symbol
Vwa5b2
Entrez Gene ID
328643
Gene Name
von Willebrand factor A domain containing 5B2
Gene Aliases
BB182430, EG328644, Gm1738, Gm931
Chromosome Location
Chr. 16: 20589319 - 20605378 on Build GRCm38
UniGene ID
Mm.40539
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001144953.1 NP_001138425.1 8 1157
NM_182636.4 NP_872574.2 8 1157
GenBank mRNA AK141799.1 - 8 1158
BC052055.1 - 7 1103

View Full Product Details