Gene Transcripts

Gene Symbol
Mettl21b
Entrez Gene ID
100504608
Gene Name
methyltransferase like 21B
Gene Aliases
Chromosome Location
Chr. 10: 127032905 - 127041394 on Build GRCm38
UniGene ID
Mm.486142
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001205036.1 NP_001191965.1 2 195

View Full Product Details