Gene Transcripts

Gene Symbol
Ech1
Entrez Gene ID
51798
Gene Name
enoyl coenzyme A hydratase 1, peroxisomal
Gene Aliases
AA617331
Chromosome Location
Chr. 7: 28824804 - 28832248 on Build GRCm38
UniGene ID
Mm.291776
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_016772.1 NP_058052.1 1 37
GenBank mRNA AF030343.1 - 1 37
AK088495.1 - 1 25
BC068112.1 - 1 37 5710971
BC087924.1 - 1 98 6706361

View Full Product Details