Gene Transcripts

Gene Symbol
Pithd1
Entrez Gene ID
66193
Gene Name
PITH (C-terminal proteasome-interacting domain of thioredoxin-like) domain containing 1
Gene Aliases
1110049F12Rik, AA408179, Trp26
Chromosome Location
Chr. 4: 135975602 - 135987244 on Build GRCm38
UniGene ID
Mm.29131
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_025411.4 NP_079687.3 2 333
GenBank mRNA AF400670.1 - 2 296
AK004208.1 - 2 334
AK050261.1 - 2 306
BC052695.1 - 2 233 5715038
BY044314.1 - 2 334

View Full Product Details