Gene Transcripts

Gene Symbol
Hddc3
Entrez Gene ID
68695
Gene Name
HD domain containing 3
Gene Aliases
1110033O09Rik, C86475, MESH1
Chromosome Location
Chr. 7: 80343083 - 80346097 on Build GRCm38
UniGene ID
Mm.21171
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_026812.2 NP_081088.1 1 61
GenBank mRNA AK004078.1 - 1 62
BC038310.1 - 1 74 5119020
BC055847.1 - 1 74

View Full Product Details