Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_019715.2 | NP_062689.2 | 3, 4 | 664 | |
GenBank mRNA | AK007070.1 | - | 3, 4 | 611 | |
AK142436.1 | - | 3, 4 | 421 | ||
AK183540.1 | - | 2, 3 | 198 | ||
BC029325.1 | - | 3, 4 | 486 | ||
BC043330.1 | - | 3, 4 | 445 | 5249829 |