Gene Transcripts

Gene Symbol
Btnl1
Entrez Gene ID
100038862
Gene Name
butyrophilin-like 1
Gene Aliases
Btnl3, Gm316, Gm33
Chromosome Location
Chr. 17: 34377123 - 34386162 on Build GRCm38
UniGene ID
Mm.440426
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001111094.1 NP_001104564.1 10 1813
GenBank mRNA BC052925.1 - 9 3255

View Full Product Details