Gene Transcripts
- Gene Symbol
- H2-Ob
- Entrez Gene ID
- 15002
- Gene Name
- histocompatibility 2, O region beta locus
- Gene Aliases
- A-beta-2, A-beta2, H-2Ab, H-2I, H-2Ob, H2-Ab, H2-Ab2, H2-IAb2, Ob
- Chromosome Location
- Chr. 17: 34238895 - 34245908 on Build GRCm38
- UniGene ID
- Mm.327358
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK053233.1 |
- |
1 |
63 |
|
AK080011.1 |
- |
1 |
221 |
|
View Full Product Details