Gene Transcripts

Gene Symbol
Zfp74
Entrez Gene ID
72723
Gene Name
zinc finger protein 74
Gene Aliases
2810054M15Rik, KRAB8, Zfp66, Znf569, mszf21, mszf77
Chromosome Location
Chr. 7: 29931711 - 29955843 on Build GRCm38
UniGene ID
Mm.371648
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
GenBank mRNA AK051279.1 - 1 212

Gene Symbol
C230062I16Rik
Entrez Gene ID
625421
Gene Name
RIKEN cDNA C230062I16 gene
Gene Aliases
Chromosome Location
Chr. 7: 29956984 - 29969764 on Build GRCm38
UniGene ID
Mm.464976
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
GenBank mRNA AK082547.1 - 232
AK138949.1 - 115

View Full Product Details