Gene Transcripts

Gene Symbol
Osbpl9
Entrez Gene ID
100273
Gene Name
oxysterol binding protein-like 9
Gene Aliases
2600011I06Rik, AU015843, Orp-9
Chromosome Location
Chr. 4: 109061145 - 109202272 on Build GRCm38
UniGene ID
Mm.366315
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001134791.2 NP_001128263.1 19 1829
NM_133885.4 NP_598646.4 22 2062
NM_173350.3 NP_775485.2 20 1868
GenBank mRNA AK035658.1 - 20 1818
AK131984.1 - 22 2034
AK172010.1 - 20 1855
BC002157.1 - 3 186
BC021507.1 - 18 1690 5347121
BC023759.1 - 22 2017
BC026927.1 - 20 1751 5134780
BC089163.1 - 19 1770 5367603

View Full Product Details