Gene Transcripts

Gene Symbol
Ric8
Entrez Gene ID
101489
Gene Name
resistance to inhibitors of cholinesterase 8 homolog (C. elegans)
Gene Aliases
AI114950, RIC-8, Ric8a
Chromosome Location
Chr. 7: 140857397 - 140863731 on Build GRCm38
UniGene ID
Mm.281916
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_053194.4 NP_444424.1 3 746
GenBank mRNA AF288813.1 - 3 745
AK031466.1 - 3 693
AK080192.1 - 3 689
AK155282.1 - 3 721
AK165536.1 - 3 723
AK167746.1 - 3 749
AK169367.1 - 3 710
AK171093.1 - 1 1038
AK187026.1 - 1 218
BC022917.1 - 3 723 4460521

View Full Product Details