Gene Transcripts

Gene Symbol
Fam208b
Entrez Gene ID
105203
Gene Name
family with sequence similarity 208, member B
Gene Aliases
AI645998, Pap20, mKIAA2006
Chromosome Location
Chr. 13: 3566035 - 3611231 on Build GRCm38
UniGene ID
Mm.221609
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_134063.3 NP_598824.3 14 4943
GenBank mRNA AK128934.1 - 1 260
AK131707.1 - 3 2339
AK165123.1 - 1 1126
AK220437.1 - 14 4931
BC085160.1 - 5 3686

View Full Product Details