Gene Transcripts

Gene Symbol
Fam19a5
Entrez Gene ID
106014
Gene Name
family with sequence similarity 19, member A5
Gene Aliases
TAFA5, Tafa-5, Tara-5
Chromosome Location
Chr. 15: 87544299 - 87759364 on Build GRCm38
UniGene ID
Mm.244319
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_134096.2 NP_598857.1 1 223
GenBank mRNA AY325124.1 - 1 224
BC015306.1 - 1 224 4223181

View Full Product Details